cis-2-(Boc-amino)cyclopropanemethanol 97%
Supplier: Thermo Scientific Chemicals
H62795-03
H62795-MD
AAH62795-03EA
706
USD
AAH62795-03
AAH62795-MD
cis-2-(Boc-amino)cyclopropanemethanol 97%
cis-2-(Boc-amino)cyclopropanemethanol
cis-2-(Boc-amino)cyclopropanemethanol, 97%
Purity: 97%
CAS No.: 170299-61-3
Molecular Formula: C9H17NO3
Formula Weight: 187.23
MDL No.: MFCD03844611
Formula:
C₉H₁₇NO₃ MW: 187.239 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03844611 CAS Number: 170299-61-3 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...