D-Alanine ethyl ester hydrochloride 95%

Supplier: Thermo Scientific Chemicals

Synonyms: H-D-Ala-OEt·HCl, H-D-Ala-OEt hydrochloride

H62791-06 H62791-14
AAH62791-06EA 42.6 USD
AAH62791-06 AAH62791-14
D-Alanine ethyl ester hydrochloride 95%
D-Alanine ethyl ester hydrochloride

D-Alanine ethyl ester hydrochloride, 95%


Purity: 95% CAS No.: 6331-09-5 Molecular Formula: C5H11NO2.HCl Formula Weight: 153.61 MDL No.: MFCD00190725

Formula: C5H12ClNO2
MW: 153.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00190725
CAS Number: 6331-09-5

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR