(S)-2-Amino-2-cyclopentylacetic acid 95%

Supplier: Thermo Scientific Chemicals

Synonyms: H-Cyclopentyl-Gly-OH, L-Cyclopentylglycine

H62776-03 H62776-06
AAH62776-03EA 243 USD
AAH62776-03 AAH62776-06 AAH62776-MD
(S)-2-Amino-2-cyclopentylacetic acid 95%
(S)-2-Amino-2-cyclopentylacetic acid

L-Cyclopentylglycine , 95%


Purity: 95% CAS No.: 2521-84-8 Molecular Formula: C7H13NO2 Formula Weight: 143.18 MDL No.: MFCD03788076

Formula: C7H13NO2
MW: 143.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03788076
CAS Number: 2521-84-8

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR