H-Orn(Boc)-OH 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
H-Orn(Boc)-OH, N-α-t-Boc-L-Ornithine, Boc-L-Orn-OH, Ndelta-Boc-L-ornithine, (S)-2-Amino-5-((tert-butoxycarbonyl)amino)pentanoic acid
H62774-03
H62774-06
AAH62774-03EA
56
USD
AAH62774-03
AAH62774-06
H-Orn(Boc)-OH 98%
H-Orn(boc)-OH
N^d-Boc-L-ornithine, 98%
Purity: 98%
CAS No.: 13650-49-2
Molecular Formula: C10H20N2O4
Formula Weight: 232.28
Storage Temperature: -30°C to -10°C
MDL No.: MFCD00037219
Formula:
C10H20N2O4 Storage Temperature: Refrigerator |
MDL Number:
MFCD00037219 CAS Number: 13650-49-2 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...