(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-(tert-butoxy)phenyl)propanoic acid 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
O-tert-Butyl-Nalpha-Fmoc-D-tyrosine, Nalpha-Fmoc-O-tert-butyl-D-tyrosine, Fmoc-D-Tyr(tBu)-OH, N-Fmoc-O-tert-butyl-D-tyrosine
H62765-03
H62765-06
H62765-14
AAH62765-03EA
57.4
USD
AAH62765-03
AAH62765-06
AAH62765-14
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-(tert-butoxy)phenyl)propanoic acid 95%
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-(tert-butoxy)phenyl)propanoic acid
N-Fmoc-O-tert-butyl-D-tyrosine, 95%
Purity: 95%
CAS No.: 118488-18-9
Molecular Formula: C28H29NO5
Formula Weight: 459.53
Storage Temperature: 2°C to 8°C
Beilstein Reference: 6691868
MDL No.: MFCD00065684
Formula:
C28H29NO5 Storage Temperature: Ambient |
MDL Number:
MFCD00065684 CAS Number: 118488-18-9 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...