N-(tert-Butoxycarbonyl)-D-glutamic acid 98%

Supplier: Thermo Scientific Chemicals

Synonyms: (R)-2-((tert-Butoxycarbonyl)amino)pentanedioic acid, Boc-D-Glu-OH, N-Boc-D-glutamic Acid

H62762-06 H62762-14
AAH62762-03EA 27.21 USD
AAH62762-03 AAH62762-06 AAH62762-14
N-(tert-Butoxycarbonyl)-D-glutamic acid 98%
N-(tert-Butoxycarbonyl)-D-glutamic acid

N-Boc-D-glutamic acid, 98%


Purity: 98% CAS No.: 34404-28-9 Molecular Formula: C10H17NO6 Formula Weight: 247.25 MDL No.: MFCD00190790

Formula: C₁₀H₁₇NO₆
MW: 247.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00190790
CAS Number: 34404-28-9

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR