cis-3-(Boc-amino)cyclohexanemethanol 97%

Supplier: Thermo Scientific Chemicals

H62744-03 H62744-MD
AAH62744-03EA 818 USD
AAH62744-03 AAH62744-MD
cis-3-(Boc-amino)cyclohexanemethanol 97%
cis-3-(Boc-amino)cyclohexanemethanol

cis-3-(Boc-amino)cyclohexanemethanol, 97%


Purity: 97% CAS No.: 920966-16-1 Molecular Formula: C12H23NO3 Formula Weight: 229.31 MDL No.: MFCD03844608

Formula: C₁₅H₂₁NO₃
Storage Temperature: Ambient
MDL Number: MFCD03844608
CAS Number: 920966-16-1

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR