trans-N-Boc-1,4-cyclohexanediamine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: trans-4-(Boc-amino)cyclohexylamine, trans-tert-Butyl (4-aminocyclohexyl)carbamate

H62734-06 H62734-14
AAH62734-06EA 130 USD
AAH62734-06 AAH62734-14
trans-N-Boc-1,4-cyclohexanediamine 97%
trans-N-Boc-1,4-cyclohexanediamine

trans-4-(Boc-amino)cyclohexylamine, 97%


Purity: 97% CAS No.: 177906-48-8 Molecular Formula: C11H22N2O2 Formula Weight: 214.30 MDL No.: MFCD03001719

Formula: C₁₁H₂₂N₂O₂
MW: 214.30 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03001719
CAS Number: 177906-48-8

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR