trans-3-(Boc-amino)cyclobutanemethanol 97%

Supplier: Thermo Scientific Chemicals

Synonyms: trans-1-(Boc-amino)-3-(hydroxymethyl)cyclobutane, tert-Butyl (trans-3-(hydroxymethyl)cyclobutyl)carbamate

H62719-03 H62719-MD
AAH62719-03EA 445 USD
AAH62719-03 AAH62719-MD
trans-3-(Boc-amino)cyclobutanemethanol 97%
trans-3-(Boc-amino)cyclobutanemethanol

trans-3-(Boc-amino)cyclobutanemethanol, 97%


Purity: 97% CAS No.: 167081-37-0 Molecular Formula: C10H19NO3 Formula Weight: 201.26 MDL No.: MFCD08166748

Formula: C₁₀H₁₉NO₃
Storage Temperature: Ambient
MDL Number: MFCD08166748
CAS Number: 167081-37-0

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR