trans-1-(Boc-amino)-3-(bromomethyl)cyclohexane 97%

Supplier: Thermo Scientific Chemicals
Warning

H62670-03 H62670-MD
AAH62670-03EA 698 USD
AAH62670-03 AAH62670-MD
trans-1-(Boc-amino)-3-(bromomethyl)cyclohexane 97%
trans-1-(Boc-amino)-3-(bromomethyl)cyclohexane

trans-1-(Boc-amino)-3-(bromomethyl)cyclohexane, 97%


Purity: 97% CAS No.: 1212405-03-2 Molecular Formula: C12H22BrNO2 Formula Weight: 292.21 MDL No.: MFCD12022626

Formula: C₁₂H₂₂BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD12022626
CAS Number: 1212405-03-2

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR