(S)-(-)-1-Methyl-2-pyrrolidinemethanol 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (S)-N-Methylpyrrolidine-2-methanol, (S)-(-)-2-Hydroxymethyl-1-methylpyrrolidine, N-Methyl-L-prolinol

H62573-06 H62573-14
AAH62573-06EA 63.7 USD
AAH62573-06 AAH62573-14
(S)-(-)-1-Methyl-2-pyrrolidinemethanol 96%
(S)-(-)-1-Methyl-2-pyrrolidinemethanol

N-Methyl-L-prolinol, 96%


Purity: 96% CAS No.: 34381-71-0 Molecular Formula: C6H13NO Formula Weight: 115.17 Flash Point: 63°(145°F) Refractive Index: 1.4690 Boiling Point: 67-69°/12mm Beilstein Reference: 79851 Optical Rotation: -49°(c=5 in methanol) MDL No.: MFCD00011727

Formula: C₆H₁₃NO
MW: 115.176 g/mol
Boiling Pt: 67…69 °C/12 mm
Density: 0.968
Flash Pt: 63 °C(145 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011727
CAS Number: 34381-71-0
EINECS: 251-981-9

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR