trans-4-(Boc-aminomethyl)cyclohexylamine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
tert-Butyl ((trans-4-aminocyclohexyl)methyl)carbamate
H62564-03
H62564-06
AAH62564-03EA
209
USD
AAH62564-03
AAH62564-06
AAH62564-MD
trans-4-(Boc-aminomethyl)cyclohexylamine 97%
trans-4-(Boc-aminomethyl)cyclohexylamine
trans-4-(Boc-aminomethyl)cyclohexylamine, 97%
Purity: 97%
CAS No.: 192323-07-2
Molecular Formula: C12H24N2O2
Formula Weight: 228.33
MDL No.: MFCD06657670
Formula:
C₁₂H₂₄N₂O₂ MW: 228.336 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06657670 CAS Number: 192323-07-2 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...