Methyl trans-4-aminocyclohexanecarboxylate hydrochloride 97%

Supplier: Thermo Scientific Chemicals

Synonyms: trans-4-Aminocyclohexanecarboxylic Acid Methyl Ester Hydrochloride

H62562-03 H62562-06
AAH62562-03EA 92.3 USD
AAH62562-03 AAH62562-06 AAH62562-MD
Methyl trans-4-aminocyclohexanecarboxylate hydrochloride 97%
Methyl trans-4-aminocyclohexanecarboxylate hydrochloride

Methyl trans-4-aminocyclohexanecarboxylate hydrochloride, 97%


Purity: 97% CAS No.: 61367-07-5 Molecular Formula: C8H15NO2.HCl Formula Weight: 193.67 MDL No.: MFCD08274538

Formula: C₈H₁₅NO₂
MW: 193.67 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08274538
CAS Number: 61367-07-5

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR