cis-4-(Benzyloxycarbonylamino)cyclohexanecarboxaldehyde 96%

Supplier: Thermo Scientific Chemicals
Warning

H62546-03 H62546-06 H62546-MD
AAH62546-03EA 145 USD
AAH62546-03 AAH62546-06 AAH62546-MD
cis-4-(Benzyloxycarbonylamino)cyclohexanecarboxaldehyde 96%
cis-4-(Benzyloxycarbonylamino)cyclohexanecarboxaldehyde

cis-4-(Benzyloxycarbonylamino)cyclohexanecarboxaldehyde, 97%


Purity: 97% CAS No.: 917022-26-5 Molecular Formula: C15H19NO3 Formula Weight: 261.31 MDL No.: MFCD03844593

Formula: C₁₅H₁₉NO₃
Storage Temperature: Ambient
MDL Number: MFCD03844593
CAS Number: 917022-26-5

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR