Boc-Trp(Boc)-OH 95%

Supplier: Thermo Scientific Chemicals

Synonyms: N-Boc-1-Boc-L-tryptophan, (S)-3-(1-(tert-Butoxycarbonyl)-1H-indol-3-yl)-2-((tert-butoxycarbonyl)amino)propanoic acid

H62542-03 H62542-06
AAH62542-03EA 42.2 USD
AAH62542-03 AAH62542-06
Boc-Trp(Boc)-OH 95%
Boc-Trp(Boc)-OH

N-Boc-1-Boc-L-tryptophan, 95%


Purity: 95% CAS No.: 144599-95-1 Molecular Formula: C21H28N2O6 Formula Weight: 404.46 Storage Temperature: -30°C to -10°C MDL No.: MFCD00270520

Formula: C₂₁H₂₈N₂O₆
MW: 404.463 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00270520
CAS Number: 144599-95-1

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR