Boc-D-Glu(Obzl)-OH 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Boc-D-Glu(OBzl)-OH, N-Boc-D-glutamic acid 5-benzyl ester

H62537-06 H62537-14
AAH62537-06EA 69.9 USD
AAH62537-06 AAH62537-14
Boc-D-Glu(Obzl)-OH 98%
Boc-D-Glu(Obzl)-OH

N-Boc-D-glutamic acid 5-benzyl ester, 98%


Purity: 98% CAS No.: 35793-73-8 Molecular Formula: C17H23NO6 Formula Weight: 337.37 MDL No.: MFCD00038258

Formula: C₁₇H₂₃NO₆
Storage Temperature: Ambient
MDL Number: MFCD00038258
CAS Number: 35793-73-8

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR