trans-3-Aminocyclohexanecarboxamide hydrochloride 97%

Supplier: Thermo Scientific Chemicals
Warning

H62529-03 H62529-MD
AAH62529-03EA 466 USD
AAH62529-03 AAH62529-MD
trans-3-Aminocyclohexanecarboxamide hydrochloride 97%
trans-3-Aminocyclohexanecarboxamide hydrochloride

trans-3-Aminocyclohexanecarboxamide hydrochloride, 97%


Purity: 97% CAS No.: 920966-29-6 Molecular Formula: C7H15ClN2O Formula Weight: 178.65 MDL No.: MFCD12022628

Formula: C₇H₁₅ClN₂O
Storage Temperature: Ambient
MDL Number: MFCD12022628
CAS Number: 920966-29-6

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR