Fmoc-4-Amino-L-phenylalanine 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Fmoc-Phe(4-NH₂)-OH, 4-Amino-N-Fmoc-L-phenylalanine, Fmoc-Phe(4-NH2)-OH, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-aminophenyl)propanoic acid, Fmoc-4-amino-Phe-OH
H62505-03
H62505-06
AAH62505-03EA
93.5
USD
AAH62505-03
AAH62505-06
AAH62505-MD
Fmoc-4-Amino-L-phenylalanine 98%
Fmoc-4-amino-L-phenylalanine
4-Amino-N-Fmoc-L-phenylalanine, 98%
Purity: 98%
CAS No.: 95753-56-3
Molecular Formula: C24H22N2O4
Formula Weight: 402.44
MDL No.: MFCD00237664
Formula:
C₂₄H₂₂N₂O₄ MW: 402.45 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00237664 CAS Number: 95753-56-3 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...