N-Fmoc-N'-trityl-D-histidine 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Fmoc-D-His(1-Trt)-OH, Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-tele-(triphenylmethyl)-D-histidine, Fmoc-D-His(Trt)-OH, (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(1-trityl-1H-imidazol-4-yl)propanoic acid, N-Fmoc-1-trityl-D-histidine, Nalpha-Fmoc-tele-trityl-D-histidine

H62504-06 H62504-14
AAH62504-06EA 101.2 USD
AAH62504-06 AAH62504-14
N-Fmoc-N'-trityl-D-histidine 98%
N-Fmoc-N'-trityl-D-histidine

N^a-Fmoc-1-trityl-D-histidine, 98%


Purity: 98% CAS No.: 135610-90-1 Molecular Formula: C40H33N3O4 Formula Weight: 619.71 Optical Rotation: -80° (c=1 in chloroform) MDL No.: MFCD00077061

Formula: C₄₀H₃₃N₃O₄
MW: 619.721 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00077061
CAS Number: 135610-90-1

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR