trans-1-(Boc-amino)-4-(bromomethyl)cyclohexane 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H62469-03
H62469-MD
AAH62469-03EA
449
USD
AAH62469-03
AAH62469-MD
trans-1-(Boc-amino)-4-(bromomethyl)cyclohexane 97%
trans-1-(Boc-amino)-4-(bromomethyl)cyclohexane
trans-1-(Boc-amino)-4-(bromomethyl)cyclohexane, 97%
Purity: 97%
CAS No.: 1222709-30-9
Molecular Formula: C12H22BrNO2
Formula Weight: 292.21
MDL No.: MFCD12827547
Formula:
C₁₂H₂₂BrNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD12827547 CAS Number: 1222709-30-9 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...