Nɑ-tert-Butoxycarbonyl-N-γ-trityl-L-asparagine 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Boc-Asn(Trt)-OH, Nɑ-Boc-Nγ-trityl-L-asparagine
H62446-03
H62446-06
AAH62446-03EA
50.8
USD
AAH62446-03
AAH62446-06
AAH62446-14
Nɑ-tert-Butoxycarbonyl-N-γ-trityl-L-asparagine 98%
Nalpha-tert-Butoxycarbonyl-N-gamma-trityl-L-asparagine
N^a-Boc-N^y-trityl-L-asparagine, 98%
Purity: 98%
CAS No.: 132388-68-2
Molecular Formula: C28H30N2O5
Formula Weight: 474.55
Melting Point: 202-205°
Beilstein Reference: 4339761
MDL No.: MFCD00153299
Formula:
C₂₈H₃₀N₂O₅ Melting Pt: 202…205 °C Storage Temperature: Ambient |
MDL Number:
MFCD00153299 CAS Number: 132388-68-2 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...