Fmoc-O-tert-butyl-D-serine 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
N-Fmoc-O-tert-butyl-D-serine, Fmoc-D-Ser(tBu)-OH
H62439-06
H62439-14
AAH62439-03EA
26.39
USD
AAH62439-03
AAH62439-06
AAH62439-14
Fmoc-O-tert-butyl-D-serine 98%
Fmoc-O-tert-butyl-D-serine
N-Fmoc-O-tert-butyl-D-serine, 98%
Purity: 98%
CAS No.: 128107-47-1
Molecular Formula: C22H25NO5
Formula Weight: 383.44
Storage Temperature: 2°C to 8°C
Melting Point: 131°
Beilstein Reference: 5309984
MDL No.: MFCD00077071
Formula:
C22H25NO5 Melting Pt: 131 °C Storage Temperature: Ambient |
MDL Number:
MFCD00077071 CAS Number: 128107-47-1 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...