N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-alanine hydrate 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
N-Fmoc-D-alanine, Fmoc-D-Ala-OH
H62395-06
H62395-14
AAH62395-06EA
90.2
USD
AAH62395-06
AAH62395-14
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-alanine hydrate 95%
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-alanine hydrate
N-Fmoc-D-alanine, 95%
Purity: 95%
CAS No.: 79990-15-1
Molecular Formula: C18H17NO4
Formula Weight: 311.34
Storage Temperature: 2°C to 8°C
Melting Point: 151-157°
Beilstein Reference: 8025133
MDL No.: MFCD00062960
Formula:
C₁₈H₁₇NO₄ MW: 311.34 g/mol Melting Pt: 151…157 °C Storage Temperature: Ambient |
MDL Number:
MFCD00062960 CAS Number: 79990-15-1 |
Specification Test Results
Assay | 95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...