(S)-Ethyl-N-Boc-pyroglutamate 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
1-Boc-L-Pyroglutamic acid ethyl ester, Boc-Pyr-OEt, Ethyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate, (S)-1-Boc-5-oxopyrrolidine-2-carboxylic acid ethyl ester, (S)-Ethyl-N-Boc-pyroglutamate, Ethyl N-(tert-Butoxycarbonyl)-L-pyroglutamate, (S)-1-(tert-Butoxycarbonyl)-5-oxopyrrolidine-2-carboxylic acid ethyl ester, Ethyl (S)-1-(tert-Butoxycarbonyl)-5-oxopyrrolidine-2-carboxylate, Ethyl N-Boc-L-pyroglutamate, N-Boc-L-pyroglutamic acid ethyl ester, N-(tert-Butoxycarbonyl)-L-pyroglutamic acid ethyl ester
Ethyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate, 98%
Purity: 98%
CAS No.: 144978-12-1
Molecular Formula: C12H19NO5
Formula Weight: 257.28
MDL No.: MFCD07778090
Formula:
C12H19NO5 Storage Temperature: Ambient |
MDL Number:
MFCD07778090 CAS Number: 144978-12-1 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...