trans-4-(Boc-amino)cyclohexaneacetaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: tert-butyl trans-N-[4-(2-oxoethyl)cyclohexyl]carbamate, tert-Butyl (trans-4-(2-oxoethyl)cyclohexyl)carbamate

H62373-03 H62373-MD
AAH62373-03EA 615 USD
AAH62373-03 AAH62373-MD
trans-4-(Boc-amino)cyclohexaneacetaldehyde 97%
trans-4-(Boc-amino)cyclohexaneacetaldehyde

trans-4-(Boc-amino)cyclohexaneacetaldehyde, 97%


Purity: 97% CAS No.: 215790-29-7 Molecular Formula: C13H23NO3 Formula Weight: 241.32 MDL No.: MFCD03844592

Formula: C₁₃H₂₃NO₃
MW: 241.331 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03844592
CAS Number: 215790-29-7

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR