Boc-L-Pro-OMe 96%
Supplier: Thermo Scientific Chemicals
Synonyms:
Boc-Pro-OMe, N-Boc-L-proline methyl ester, N-(tert-Butoxycarbonyl)-L-proline methyll ester
H62363-14
H62363-22
AAH62363-14EA
71.7
USD
AAH62363-14
AAH62363-22
Boc-L-Pro-OMe 96%
Boc-L-Pro-OMe
N-Boc-L-proline methyl ester, 96%
Purity: 96%
CAS No.: 59936-29-7
Molecular Formula: C11H19NO4
Formula Weight: 229.27
MDL No.: MFCD01075092
Formula:
C11H19NO4 Storage Temperature: Ambient |
MDL Number:
MFCD01075092 CAS Number: 59936-29-7 |
Specification Test Results
Assay | 96% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...