(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-aspartic Acid 4-tert-Butyl Ester, N-Fmoc-D-aspartic Acid 4-tert-Butyl Ester, 4-tert-Butyl N-Fmoc-D-aspartate, Fmoc-D-Asp(OtBu)-OH
H62356-03
H62356-06
AAH62356-03EA
72.1
USD
AAH62356-03
AAH62356-06
AAH62356-MD
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid 98%
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid
N-Fmoc-D-aspartic acid 4-tert-butyl ester, 98%
Purity: 98%
CAS No.: 112883-39-3
Molecular Formula: C23H25NO6
Formula Weight: 411.45
MDL No.: MFCD00077050
Formula:
C23H25NO6 Storage Temperature: Ambient |
MDL Number:
MFCD00077050 CAS Number: 112883-39-3 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...