(S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid 95%

Supplier: Thermo Scientific Chemicals

Synonyms: N-Fmoc-L-aspartic Acid 1-tert-Butyl Ester, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartic Acid 1-tert-Butyl Ester, Fmoc-Asp(OH)-OtBu, Fmoc-Asp-OtBu, 1-tert-Butyl N-Fmoc-L-aspartate

H62355-03 H62355-06
AAH62355-03EA 35 USD
AAH62355-03 AAH62355-06
(S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid 95%
(S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid

N-Fmoc-L-aspartic acid 1-tert-butyl ester, 95%


Purity: 95% CAS No.: 129460-09-9 Molecular Formula: C23H25NO6 Formula Weight: 411.45 Beilstein Reference: 3658045 MDL No.: MFCD00065631

Formula: C23H25NO6
MW: 411.5 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00065631
CAS Number: 129460-09-9

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR