N-Boc-DL-phenylglycine 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Boc-DL-Phg-OH

H62354-06 H62354-14
AAH62354-06EA 58 USD
AAH62354-06 AAH62354-14
N-Boc-DL-phenylglycine 98%
N-Boc-DL-phenylglycine

N-Boc-DL-phenylglycine, 98%


Purity: 98% CAS No.: 3601-66-9 Molecular Formula: C13H17NO4 Formula Weight: 251.28 MDL No.: MFCD00971063

Formula: C₁₃H₁₇NO₄
MW: 251.282 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00971063
CAS Number: 3601-66-9

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR