cis-2-(Benzyloxycarbonylamino)cyclobutanecarboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

H62329-03 H62329-MD
AAH62329-03EA 1204 USD
AAH62329-03 AAH62329-MD
cis-2-(Benzyloxycarbonylamino)cyclobutanecarboxylic acid 97%
cis-2-(Benzyloxycarbonylamino)cyclobutanecarboxylic acid

cis-2-(Benzyloxycarbonylamino)cyclobutanecarboxylic acid, 97%


Purity: 97% CAS No.: 685508-28-5 Molecular Formula: C13H15NO4 Formula Weight: 249.26 MDL No.: MFCD03844615

Formula: C₁₃H₁₅NO₄
MW: 249.266 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03844615
CAS Number: 685508-28-5

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR