Pyrrolidin-2-ylmethanol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: DL-Prolinol, 2-Pyrrolidinemethanol, 2-(Hydroxymethyl)pyrrolidine

H62322-06 H62322-14
AAH62322-06EA 64.2 USD
AAH62322-06 AAH62322-14
Pyrrolidin-2-ylmethanol 98%
Pyrrolidin-2-ylmethanol

DL-Prolinol, 98%


Purity: 98% CAS No.: 498-63-5 Molecular Formula: C5H11NO Formula Weight: 101.15 Refractive Index: 1.4840 Boiling Point: 76°/2mm MDL No.: MFCD00601073

Formula: C5H11NO
MW: 101.15 g/mol
Boiling Pt: 76 °C/2 mm
Storage Temperature: Ambient
MDL Number: MFCD00601073
CAS Number: 498-63-5

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR