Z-Asp-OBzl 95%

Supplier: Thermo Scientific Chemicals

Synonyms: N-Benzyloxycarbonyl-L-aspartic acid 1-benzyl ester, Cbz-Asp-OBzl

H62306-03 H62306-06 H62306-14
AAH62306-03EA 33.7 USD
AAH62306-03 AAH62306-06 AAH62306-14
Z-Asp-OBzl 95%
Z-Asp-OBzl

N-Benzyloxycarbonyl-L-aspartic acid 1-benzyl ester, 95%


Purity: 95% CAS No.: 4779-31-1 Molecular Formula: C19H19NO6 Formula Weight: 357.36 MDL No.: MFCD00038594

Formula: C₁₉H₁₉NO₆
MW: 357.362 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00038594
CAS Number: 4779-31-1

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR