Ethyl-1-(hydroxymethyl)cyclopropanecarboxylate 95%

Supplier: Thermo Scientific Chemicals

Synonyms: Ethyl 1-(hydroxymethyl)cyclopropanecarboxylate

H62277-03 H62277-06 H62277-MD
AAH62277-03EA 236 USD
AAH62277-03 AAH62277-06 AAH62277-MD
Ethyl-1-(hydroxymethyl)cyclopropanecarboxylate 95%
Ethyl-1-(hydroxymethyl)cyclopropanecarboxylate

Ethyl 1-(hydroxymethyl)cyclopropanecarboxylate acts as a ligand for Suzuki coupling reaction.


Purity: 97% CAS No.: 3697-68-5 Molecular Formula: C7H12O3 Formula Weight: 144.16 MDL No.: MFCD09759206

Formula: C₇H₁₂O₃
MW: 144.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09759206
CAS Number: 3697-68-5

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR