Octenidine dihydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,1'-(Decane-1,10-diyl)bis(N-octylpyridin-4(1H)-imine) dihydrochloride

H62257-03 H62257-06
AAH62257-03EA 38.4 USD
AAH62257-03 AAH62257-06
Octenidine dihydrochloride 98%
Octenidine dihydrochloride

Octenidine dihydrochloride, 98%


Purity: 98% CAS No.: 70775-75-6 Molecular Formula: C36H64Cl2N4 Formula Weight: 623.83 Melting Point: 214° MDL No.: MFCD01938808

Formula: C₃₆H₆₂N₄
MW: 623.84 g/mol
Melting Pt: 214 °C
Storage Temperature: Ambient
MDL Number: MFCD01938808
CAS Number: 70775-75-6
EINECS: 274-861-8
Merck Index: 14,6754

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR