H-Val-OBzl·HCl 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
L-Valine Benzyl Ester Hydrochloride, (S)-benzyl 2-amino-3-methylbutanoate hydrochloride, benzyl (2S)-2-amino-3-methylbutanoate hydrochloride, H-Val-Obzl.HCl
H62242-06
H62242-14
AAH62242-06EA
55.1
USD
AAH62242-06
AAH62242-14
H-Val-OBzl·HCl 95%
H-Val-OBzl·HCl
L-Valine benzyl ester hydrochloride, 95%
Purity: 95%
CAS No.: 2462-34-2
Molecular Formula: C12H17NO2.HCl
Formula Weight: 243.73
Storage Temperature: -30°C to -10°C
Melting Point: 138°
Optical Rotation: -10°(c=2 in water)
MDL No.: MFCD00058010
Formula:
C₁₂H₁₈ClNO₂ MW: 243.731 g/mol Melting Pt: 138 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00058010 CAS Number: 2462-34-2 |
Specification Test Results
Assay | 95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...