cis-4-(Boc-amino)cyclohexanol 97%

Supplier: Thermo Scientific Chemicals
Warning

H62238-03 H62238-06 H62238-MD
AAH62238-03EA 155 USD
AAH62238-03 AAH62238-06 AAH62238-MD
cis-4-(Boc-amino)cyclohexanol 97%
cis-4-(Boc-amino)cyclohexanol

cis-4-(Boc-amino)cyclohexanol, 97%


Purity: 97% CAS No.: 167081-25-6 Molecular Formula: C11H21NO3 Formula Weight: 215.28 MDL No.: MFCD03844614

Formula: C₁₁H₂₁NO₃
Storage Temperature: Ambient
MDL Number: MFCD03844614
CAS Number: 167081-25-6

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR