trans-2-(Boc-amino)cyclopropanemethanol 97%

Supplier: Thermo Scientific Chemicals

Synonyms: rel-tert-Butyl ((1R,2R)-2-(hydroxymethyl)cyclopropyl)carbamate

H62222-03 H62222-MD
AAH62222-03EA 557 USD
AAH62222-03 AAH62222-MD
trans-2-(Boc-amino)cyclopropanemethanol 97%
trans-2-(Boc-amino)cyclopropanemethanol

trans-2-(Boc-amino)cyclopropanemethanol, 97%


Purity: 97% CAS No.: 170299-53-3 Molecular Formula: C9H17NO3 Formula Weight: 187.23 MDL No.: MFCD03844612

Formula: C₉H₁₇NO₃
Storage Temperature: Ambient
MDL Number: MFCD03844612
CAS Number: 170299-53-3

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR