N-Boc-3,5-difluoro-L-phenylalanine 95%

Supplier: Thermo Scientific Chemicals

Synonyms: Boc-Phe(3,5-DiF)-OH

H62216-03 H62216-06 H62216-MD
AAH62216-03EA 97.4 USD
AAH62216-03 AAH62216-06 AAH62216-MD
N-Boc-3,5-difluoro-L-phenylalanine 95%
N-Boc-3,5-difluoro-L-phenylalanine

N-Boc-3,5-difluoro-L-phenylalanine, 95%


Purity: 95% CAS No.: 205445-52-9 Molecular Formula: C14H17F2NO4 Formula Weight: 301.29 Melting Point: 115° MDL No.: MFCD00797555

Formula: C₁₄H₁₇F₂NO₄
MW: 301.29 g/mol
Melting Pt: 115 °C
Storage Temperature: Ambient
MDL Number: MFCD00797555
CAS Number: 205445-52-9

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR