cis-4-(Boc-amino)cyclohexanemethylamine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
tert-Butyl (cis-4-(aminomethyl)cyclohexyl)carbamate
H62187-03
H62187-MD
AAH62187-03EA
406
USD
AAH62187-03
AAH62187-MD
cis-4-(Boc-amino)cyclohexanemethylamine 97%
cis-4-(Boc-amino)cyclohexanemethylamine
cis-4-(Boc-amino)cyclohexanemethylamine, 97%
Purity: 97%
CAS No.: 509143-00-4
Molecular Formula: C12H24N2O2
Formula Weight: 228.33
MDL No.: MFCD03844602
Formula:
C₁₂H₂₄N₂O₂ MW: 228.336 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03844602 CAS Number: 509143-00-4 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...