Fmoc-D-Trp(Boc)-OH 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 1-Boc-N-Fmoc-D-tryptophan

H62185-03 H62185-06
AAH62185-03EA 74.9 USD
AAH62185-03 AAH62185-06 AAH62185-MD
Fmoc-D-Trp(Boc)-OH 98%
Fmoc-D-Trp(Boc)-OH

1-Boc-N-Fmoc-D-tryptophan, 98%


Purity: 98% CAS No.: 163619-04-3 Molecular Formula: C31H30N2O6 Formula Weight: 526.59 Storage Temperature: 2°C to 8°C Beilstein Reference: 7062970 MDL No.: MFCD00153367

Formula: C₃₁H₃₀N₂O₆
MW: 526.589 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00153367
CAS Number: 163619-04-3

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR