Boc-Arg(Pbf)-OH 96%
Supplier: Thermo Scientific Chemicals
Synonyms:
(S)-2-((tert-Butoxycarbonyl)amino)-5-(3-((2,2,4,6,7-pentamethyl-2,3-dihydrobenzofuran-5-yl)sulphonyl)guanidino)pentanoic acid, Nα-Boc-Nomega-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-sulphonyl)-L-arginine
H62148-03
H62148-06
AAH62148-03EA
62.1
USD
AAH62148-03
AAH62148-06
Boc-Arg(Pbf)-OH 96%
Boc-Arg(Pbf)-OH
N^a-Boc-N^w-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-sulfonyl)-L-arginine, 96%
Purity: 96%
CAS No.: 200124-22-7
Molecular Formula: C24H38N4O7S
Formula Weight: 526.65
Storage Temperature: 2°C to 8°C
MDL No.: MFCD00236821
Formula:
C₂₄H₃₈N₄O₇S Storage Temperature: Ambient |
MDL Number:
MFCD00236821 CAS Number: 200124-22-7 |
Specification Test Results
Assay | 96% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...