Boc-Ser(tBu)-OH 98%
Supplier: Thermo Scientific Chemicals
H62147-03
H62147-06
AAH62147-03EA
57.4
USD
AAH62147-03
AAH62147-06
Boc-Ser(tBu)-OH 98%
Boc-Ser(tBu)-OH
N-Boc-O-tert-butyl-L-serine, 98%
Purity: 98%
CAS No.: 13734-38-8
Molecular Formula: C12H23NO5
Formula Weight: 261.31
MDL No.: MFCD00079666
Formula:
C12H23NO5 Storage Temperature: Ambient |
MDL Number:
MFCD00079666 CAS Number: 13734-38-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...