Fmoc-Glu-OtBu 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
(S)-4-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-(tert-butoxy)-5-oxopentanoic acid, N-Fmoc-L-glutamic acid 1-tert-butyl ester, Fmoc-Glu-OtBu
H62146-03
H62146-06
AAH62146-03EA
124
USD
AAH62146-03
AAH62146-06
AAH62146-MD
Fmoc-Glu-OtBu 98%
Fmoc-Glu-OtBu
N-Fmoc-L-glutamic acid 1-tert-butyl ester, 98%
Purity: 98%
CAS No.: 84793-07-7
Molecular Formula: C24H27NO6
Formula Weight: 425.47
MDL No.: MFCD00065648
Formula:
C₂₄H₂₇NO₆ MW: 425.47 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00065648 CAS Number: 84793-07-7 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...