2-(((Benzyloxy)carbonyl)(methyl)amino)acetic acid 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
N-Cbz-N-methylglycine, Z-N-Me-Gly-OH, Z-Sar-OH, N-((Benzyloxy)carbonyl)-N-methylglycine, N-(Benzyloxycarbonyl)sarcosine
H62135-06
H62135-14
AAH62135-06EA
41.5
USD
AAH62135-06
AAH62135-14
2-(((Benzyloxy)carbonyl)(methyl)amino)acetic acid 95%
2-(((Benzyloxy)carbonyl)(methyl)amino)acetic acid
N-(Benzyloxycarbonyl)sarcosine, 95%
Purity: 95%
CAS No.: 39608-31-6
Molecular Formula: C11H13NO4
Formula Weight: 223.23
Storage Temperature: -30°C to -10°C
MDL No.: MFCD00021747
Formula:
C₁₁H₁₃NO₄ MW: 223.3 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00021747 CAS Number: 39608-31-6 |
Specification Test Results
Assay | 95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...