Dimethyl-D-glutamate hydrochloride 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
H-D-Glu(OMe)-OMe.HCl, H-D-Glu(OMe)-OMe·HCl, D-Glutamic Acid Dimethyl Ester Hydrochloride
H62132-03
H62132-06
H62132-14
AAH62132-03EA
25
USD
AAH62132-03
AAH62132-06
AAH62132-14
Dimethyl-D-glutamate hydrochloride 98%
Dimethyl-D-glutamate hydrochloride
D-Glutamic acid dimethyl ester hydrochloride, 98%
Purity: 98%
CAS No.: 27025-25-8
Molecular Formula: C7H13NO4.HCl
Formula Weight: 211.64
Storage Temperature: 2°C to 8°C
Melting Point: 91°
Optical Rotation: -26°(c=5 in water)
MDL No.: MFCD00235872
Formula:
C₇H₁₃NO₄ MW: 211.64 g/mol Melting Pt: 91 °C Storage Temperature: Ambient |
MDL Number:
MFCD00235872 CAS Number: 27025-25-8 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...