H-D-Val-OtBu·HCl 95%
Supplier: Thermo Scientific Chemicals
H62131-03
H62131-06
H62131-14
AAH62131-03EA
36.6
USD
AAH62131-03
AAH62131-06
AAH62131-14
H-D-Val-OtBu·HCl 95%
H-D-Val-OtBu·HCl
D-Valine tert-butyl ester hydrochloride, 95%
Purity: 95%
CAS No.: 104944-18-5
Molecular Formula: C9H19NO2.HCl
Formula Weight: 209.71
Storage Temperature: -30°C to -10°C
MDL No.: MFCD03939779
Formula:
C₉H₂₀ClNO₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD00237308 CAS Number: 104944-18-5 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...