O-tert-Butyl-L-serine Methyl Ester Hydrochloride 98%

Supplier: Thermo Scientific Chemicals

Synonyms: H-Ser(tBu)-OMe·HCl, H-Ser(tBu)-OMe.HCl

H62126-03 H62126-06 H62126-MD
AAH62126-03EA 76.3 USD
AAH62126-03 AAH62126-06 AAH62126-MD
O-tert-Butyl-L-serine Methyl Ester Hydrochloride 98%
O-tert-Butyl-L-serine methyl ester hydrochloride

O-tert-Butyl-L-serine methyl ester hydrochloride, 98%


Purity: 98% CAS No.: 17114-97-5 Molecular Formula: C8H17NO3.HCl Formula Weight: 211.69 Beilstein Reference: 2041979 MDL No.: MFCD00077108

Formula: C₈H₁₇NO₃
MW: 211.69 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00077108
CAS Number: 17114-97-5

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR