Methyl-L-threoninate hydrochloride 98%

Supplier: Thermo Scientific Chemicals

Synonyms: (2S,3R)-Methyl 2-amino-3-hydroxybutanoate hydrochloride, H-Thr-OMe·HCl, L-Threonine methyl ester hydrochloride

H62118-03 H62118-06 H62118-14
AAH62118-03EA 52.8 USD
AAH62118-03 AAH62118-06 AAH62118-14
Methyl-L-threoninate hydrochloride 98%
Methyl-L-threoninate hydrochloride

L-Threonine methyl ester hydrochloride, 98%


Purity: 98% CAS No.: 39994-75-7 Molecular Formula: C5H11NO3.HCl Formula Weight: 169.61 Storage Temperature: -30°C to -10°C Beilstein Reference: 3562717 MDL No.: MFCD00037677

Formula: C₅H₁₂ClNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD00037677
CAS Number: 39994-75-7
EINECS: 254-738-5

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR