Boc-D-Lys(fmoc)-OH 95%

Supplier: Thermo Scientific Chemicals

Synonyms: Nα-Boc-Nepsilon-Fmoc-D-lysine, Boc-D-Lys(Fmoc)-OH

H62116-03 H62116-06
AAH62116-03EA 122 USD
AAH62116-03 AAH62116-06 AAH62116-MD
Boc-D-Lys(fmoc)-OH 95%
Boc-D-Lys(fmoc)-OH

N^a-Boc-N^e-Fmoc-D-lysine, 95%


Purity: 95% CAS No.: 115186-31-7 Molecular Formula: C26H32N2O6 Formula Weight: 468.55 Melting Point: 117-123° MDL No.: MFCD00076966

Formula: C₂₆H₃₂N₂O₆
MW: 468.55 g/mol
Melting Pt: 117…123 °C
Storage Temperature: Ambient
MDL Number: MFCD00076966
CAS Number: 115186-31-7

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR