Boc-L-pyroglutamic acid methyl ester 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl (S)-1-(tert-Butoxycarbonyl)-5-oxopyrrolidine-2-carboxylate, Methyl N-Boc-L-pyroglutamate, Methyl N-(tert-Butoxycarbonyl)-L-pyroglutamate, Boc-Pyr-OMe, (S)-1-(tert-Butoxycarbonyl)-5-oxopyrrolidine-2-carboxylic acid methyll ester, N-Boc-L-pyroglutamic acid methyll ester, (S)-1-Boc-5-oxopyrrolidine-2-carboxylic acid methyll ester, N-(tert-Butoxycarbonyl)-L-pyroglutamic acid methyll ester, 1-(tert-Butyl) 2-methyl (S)-5-oxopyrrolidine-1,2-dicarboxylate, Methyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate

H62091-03 H62091-06 H62091-14
AAH62091-03EA 27.6 USD
AAH62091-03 AAH62091-06 AAH62091-14
Boc-L-pyroglutamic acid methyl ester 98%
Boc-L-pyroglutamic acid methyl ester

Methyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate, 98%


Purity: 98% CAS No.: 108963-96-8 Molecular Formula: C11H17NO5 Formula Weight: 243.26 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White Melting Point: 68-72° Optical Rotation: -32°(c=0.5 in chloroform) UN No.: 3077 MDL No.: MFCD06809720

Formula: C11H17NO5
MW: 243.26 g/mol
Melting Pt: 68…72 °C
Storage Temperature: Ambient
MDL Number: MFCD06809720
CAS Number: 108963-96-8
UN: 3077
ADR: "9",III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR